missing translation for 'onlineSavingsMsg'
Learn More

4-Trimethylsilyl-3-butyn-2-ol, 97%

Product Code. 11323696
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11323696 5 g 5g
11333696 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11323696 Supplier Thermo Scientific Alfa Aesar Supplier No. L14815.06
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6999-19-5 | C7H14OSi | 142.273 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6999-19-5
Molecular Formula C7H14OSi
Molecular Weight (g/mol) 142.273
MDL Number MFCD00190213
InChI Key HJJSDJHRTMFJLP-UHFFFAOYSA-N
Synonym 4-trimethylsilyl-3-butyn-2-ol, 4-trimethylsilyl but-3-yn-2-ol, 1-trimethylsilylbut-1-yne-3-ol, +/-4-trimethylsilyl-3-butyn-2-ol, acmc-20mpjc, 3-butyn-2-ol, 4-trimethylsilyl-, 2s, acmc-1bfey, hjjsdjhrtmfjlp-uhfffaoysa, 3-butyn-2-ol,4-trimethylsilyl
PubChem CID 2760828
IUPAC Name 4-trimethylsilylbut-3-yn-2-ol
SMILES CC(C#C[Si](C)(C)C)O

Specifications

Boiling Point 126°C to 129°C
Quantity 5 g
Flash Point 57°C (135°F)
Refractive Index 1.446
Density 0.846
UN Number UN1987
Beilstein 1923632
Formula Weight 142.27
Percent Purity 97%
Chemical Name or Material 4-Trimethylsilyl-3-butyn-2-ol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.