missing translation for 'onlineSavingsMsg'
Learn More

4-Trimethylsilyl-3-butyn-1-ol, 98%

Product Code. 13498528 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
13498528 25 g Glass bottle 25g
10065463 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 13498528 Supplier Thermo Scientific Acros Supplier No. 381320250
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2117-12-6 | C7H14OSi | 142.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2117-12-6
Molecular Formula C7H14OSi
Molecular Weight (g/mol) 142.27
MDL Number MFCD00190217
InChI Key VSTMVAFCUIGHQI-UHFFFAOYSA-N
Synonym 4-trimethylsilyl-3-butyn-1-ol, 4-trimethylsilyl but-3-yn-1-ol, 3-butyn-1-ol, 4-trimethylsilyl, 4-trimethylsilyl-3-butyne-1-ol, 1-trimethylsilyl-1-butyn-4-ol, 3-butyn-1-ol,4-trimethylsilyl
PubChem CID 2760827
IUPAC Name 4-trimethylsilylbut-3-yn-1-ol
SMILES C[Si](C)(C)C#CCCO

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Black-Brown to Gray
Density 0.8540g/mL
Boiling Point 97.0°C to 98.0°C (24.0 mmHg)
Flash Point 60°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3SiC≡C(CH2)2OH
Refractive Index 1.4540 to 1.4560 (20°C, 589nm)
Quantity 25 g
Specific Gravity 0.854
Formula Weight 142.27
Percent Purity 98%
Physical Form Solution
Chemical Name or Material 4-Trimethylsilyl-3-butyn-1-ol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-Trimethylsilyl-3-butyn-1-ol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.