missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethylthio)phenyl isocyanate, 97%, Thermo Scientific™

Product Code. 11324076
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 11324076

Brand: Thermo Scientific Alfa Aesar L19206.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Solubility
It hydrolyzes with water.

Notes
Moisture sensitive. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24032-84-6
Molecular Formula C8H4F3NOS
Molecular Weight (g/mol) 219.181
MDL Number MFCD00036025
InChI Key NYQSIHZNEJCZKX-UHFFFAOYSA-N
Synonym 4-trifluoromethylthio phenyl isocyanate, 1-isocyanato-4-trifluoromethyl sulfanyl benzene, 4-trifluoromethylthio phenylisocyanate, 1-isocyanato-4-trifluoromethylsulfanyl benzene, 1-isocyanato-4-trifluoromethyl thio benzene, acmc-20amtf, pubchem5000, timtec-bb sbb006639, 4-trifluoromethylthio benzenisocyanate, 4-trifluoromethyl thiophenyl isocyanate
PubChem CID 2733392
IUPAC Name 1-isocyanato-4-(trifluoromethylsulfanyl)benzene
SMILES C1=CC(=CC=C1N=C=O)SC(F)(F)F

Specifications

Density 1.385
Boiling Point 175°C
Flash Point 80°C (176°F)
Refractive Index 1.511
Quantity 1 g
UN Number UN2206
Beilstein 6290747
Sensitivity Moisture sensitive
Solubility Information It hydrolyzes with water.
Formula Weight 219.19
Percent Purity 97%
Chemical Name or Material 4-(Trifluoromethylthio)phenyl isocyanate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.