missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethylthio)benzyl bromide, 97%, Thermo Scientific™

Product Code. 11304076
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 11304076

Brand: Thermo Scientific Alfa Aesar L19205.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21101-63-3
Molecular Formula C8H6BrF3S
Molecular Weight (g/mol) 271.095
MDL Number MFCD00129189
InChI Key VAYGJBCOCRVROJ-UHFFFAOYSA-N
Synonym 4-trifluoromethylthio benzyl bromide, 4-bromomethyl phenyl trifluoromethyl sulfane, 4-trifluoromethylthio benzylbromide, 1-bromomethyl-4-trifluoromethyl sulfanyl benzene, benzene, 1-bromomethyl-4-trifluoromethyl thio, 1-bromomethyl-4-trifluoromethylsulfanyl-benzene, 1-bromomethyl-4-trifluoromethylsulfanyl benzene, acmc-1coe0, 4-trifluoromethylthiobenzyl bromide, 4 trifluoromethylthio benzyl bromide
PubChem CID 2777882
IUPAC Name 1-(bromomethyl)-4-(trifluoromethylsulfanyl)benzene
SMILES C1=CC(=CC=C1CBr)SC(F)(F)F

Specifications

Melting Point 53°C to 56°C
Boiling Point 115°C to 118°C (13 mmHg)
Odor Pungent
Quantity 5 g
UN Number UN3261
Beilstein 2209970
Formula Weight 271.1
Percent Purity 97%
Chemical Name or Material 4-(Trifluoromethylthio)benzyl bromide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.