missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethylthio)benzaldehyde, 90+%

Product Code. 11469826
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11469826

Brand: Thermo Scientific Alfa Aesar B20313.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4021-50-5 | C8H5F3OS | 206.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4021-50-5
Molecular Formula C8H5F3OS
Molecular Weight (g/mol) 206.18
MDL Number MFCD00129191
InChI Key OPAXHVNZGUGCKW-UHFFFAOYSA-N
Synonym 4-trifluoromethylthio benzaldehyde, 4-trifluoromethyl thio benzaldehyde, 4-trifluoromethyl sulfanyl benzaldehyde, benzaldehyde, 4-trifluoromethyl thio, acmc-1agrc, ksc582m6p, 4-trifluomethylthio benzoldehyde, rarechem gt hw 0381
PubChem CID 2777846
IUPAC Name 4-(trifluoromethylsulfanyl)benzaldehyde
SMILES FC(F)(F)SC1=CC=C(C=O)C=C1

Specifications

Melting Point 29°C to 31°C
Density 1.31
Boiling Point 219°C
Flash Point 96°C (204°F)
Odor Stench
Refractive Index 1.511
Quantity 1 g
Beilstein 2328411
Sensitivity Air Sensitive
Formula Weight 206.19
Percent Purity ≥90%
Chemical Name or Material 4-(Trifluoromethylthio)benzaldehyde
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.