missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethylsulfonyl)phenylacetic acid, 98%

Product Code. 15475007
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15475007 1 g 1g
15485007 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15475007 Supplier Thermo Scientific Alfa Aesar Supplier No. H32295.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1099597-82-6 | C9H7F3O4S | 268.206 g/mol

4-(Trifluoromethylsulfonyl)phenylacetic acid, is used as a primary and secondary intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Trifluoromethylsulfonyl)phenylacetic acid, is used as a primary and secondary intermediate.

Solubility
Slightly soluble in water, soluble in organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1099597-82-6
Molecular Formula C9H7F3O4S
Molecular Weight (g/mol) 268.206
MDL Number MFCD04973012
InChI Key FIEUCHICQSJBAU-UHFFFAOYSA-N
Synonym 4-trifluoromethylsulfony phenylacetic acid, 2-4-trifluoromethyl sulfonyl phenyl acetic acid, 4-trifluoromethanesulfonylphenyl acetic acid, 4-trifluoromethylsulfonyl phenylacetic acid, 4-trifluoromethanesulfonyl phenyl acetic acid
PubChem CID 40427164
IUPAC Name 2-[4-(trifluoromethylsulfonyl)phenyl]acetic acid
SMILES C1=CC(=CC=C1CC(=O)O)S(=O)(=O)C(F)(F)F

Specifications

Melting Point 125°C to 128°C
Quantity 1 g
Solubility Information Slightly soluble in water,soluble in organic solvents.
Formula Weight 268.21
Percent Purity 98%
Chemical Name or Material 4-(Trifluoromethylsulfonyl)phenylacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.