missing translation for 'onlineSavingsMsg'
Learn More

4-Trifluoromethylphenylboronic acid, 98%, Thermo Scientific Chemicals

Product Code. 10304012 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mg
5 g
Packaging:
Glass bottle
Unit Size:
500mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10304012 500 mg Glass bottle 500mg
10360144 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10304012 Supplier Thermo Scientific Acros Supplier No. 359765000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 128796-39-4
Molecular Formula C7H6BF3O2
Molecular Weight (g/mol) 189.93
MDL Number MFCD00151855
InChI Key ALMFIOZYDASRRC-UHFFFAOYSA-N
Synonym 4-trifluoromethyl phenylboronic acid, 4-trifluoromethylphenylboronic acid, 4-trifluoromethyl benzeneboronic acid, 4-trifluoromethyl phenyl boronic acid, 4-trifluoromethylphenyl boronic acid, 4-trifluoromethyl phenyl boranediol, 4-boronobenzotrifluoride, p-trifluoromethyl phenylboronic acid, 4-trifluoromethylphenyl-boronic acid
PubChem CID 2734389
IUPAC Name [4-(trifluoromethyl)phenyl]boronic acid
SMILES OB(O)C1=CC=C(C=C1)C(F)(F)F

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 245.0°C to 250.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 500 mg
Formula Weight 189.93
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 4-Trifluoromethylphenylboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 4-Trifluoromethylphenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.