missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)thiophenol, 97%

Product Code. 11410673
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11410673 5 g 5g
11400673 1 g 1g
11420673 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11410673 Supplier Thermo Scientific Alfa Aesar Supplier No. A12725.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 825-83-2 | C7H5F3S | 178.172 g/mol

4-(Trifluoromethyl)thiophenol is used an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Trifluoromethyl)thiophenol is used an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals.

Solubility
Insoluble in water.

Notes
Air Sensitive. Store in cool dry place in tightly closed container. With good ventilation. Store away from air and oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 825-83-2
Molecular Formula C7H5F3S
Molecular Weight (g/mol) 178.172
MDL Number MFCD00042320
InChI Key WCMLRSZJUIKVCW-UHFFFAOYSA-N
Synonym 4-trifluoromethyl thiophenol, 4-trifluoromethyl benzenethiol, 4-trifluoromethyl benzene-1-thiol, 4-trifluoromethylthiophenol, 4-mercaptobenzotrifluoride, benzenethiol, 4-trifluoromethyl, 4-trifluoromethyl t, acmc-20amtl, pubchem6864
PubChem CID 136653
IUPAC Name 4-(trifluoromethyl)benzenethiol
SMILES C1=CC(=CC=C1C(F)(F)F)S

Specifications

Density 1.3
Boiling Point 66°C to 68°C (15 mmHg)
Odor Stench
Refractive Index 1.492
Quantity 5 g
UN Number UN2810
Beilstein 2045665
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 178.18
Percent Purity 97%
Chemical Name or Material 4-(Trifluoromethyl)thiophenol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.