missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)pyridine-3-carbothioamide, ≥95%, Thermo Scientific™

Product Code. 10546435
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10546435 5 g 5g
10029591 250 mg 250mg
10561893 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10546435 Supplier Thermo Scientific Maybridge Supplier No. SPB05766DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 158063-54-8
Molecular Formula C7H5F3N2S
Molecular Weight (g/mol) 206.186
MDL Number MFCD00205806
InChI Key HOPAEPDXFOSQMZ-UHFFFAOYSA-N
PubChem CID 2777771
IUPAC Name 4-(trifluoromethyl)pyridine-3-carbothioamide
SMILES C1=CN=CC(=C1C(F)(F)F)C(=S)N

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Quantity 5 g
Synonym 4-trifluoromethyl pyridine-3-carbothioamide, 4-trifluoromethyl-3-pyridinecarbothioamide, 4-trifluoromethyl nicotinthioamide, 4-trifluoromethyl pyridine-3-thiocarboxamide, maybridge4_004528, 3-pyridinecarbothioamide, 4-trifluoromethyl, 4-trifluoromethyl-thionicotinamide, 4-trifluoromethylpyridine-3-thiocarboxamide, 3-pyridinecarbothioamide,4-trifluoromethyl, amino 4-trifluoromethyl 3-pyridyl methane-1-thione
Formula Weight 206.19
Percent Purity 95%
Chemical Name or Material 4-(Trifluoromethyl)pyridine-3-carbothioamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.