missing translation for 'onlineSavingsMsg'
Learn More

[4-(Trifluoromethyl)phenylthio]acetic acid, Thermo Scientific™

Product Code. 11439420
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11439420

Brand: Thermo Scientific Alfa Aesar H50283.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102582-93-4
Molecular Formula C9H7F3O2S
Molecular Weight (g/mol) 236.208
MDL Number MFCD00236351
InChI Key MBVVWIKWEHKYGF-UHFFFAOYSA-N
Synonym 2-4-trifluoromethyl phenyl thio acetic acid, 2-4-trifluoromethyl phenylthio acetic acid, 4-trifluoromethyl phenyl sulfanyl acetic acid, 2-4-trifluoromethyl phenyl sulfanylacetic acid, acetic acid, 4-trifluoromethyl phenyl thio, acetic acid, 2-4-trifluoromethyl phenyl thio, cbmicro_022213, acmc-1cqmh, 4-carboxymethyl thio benzotrifluoride
PubChem CID 5339850
IUPAC Name 2-[4-(trifluoromethyl)phenyl]sulfanylacetic acid
SMILES C1=CC(=CC=C1C(F)(F)F)SCC(=O)O

Specifications

Melting Point 114°C to 118°C
Quantity 1 g
Formula Weight 236.21
Chemical Name or Material [4-(Trifluoromethyl)phenylthio]acetic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.