missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)phenylacetonitrile, 98%, Thermo Scientific™

Product Code. 11413987
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11413987 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11413987 Supplier Thermo Scientific Alfa Aesar Supplier No. A19386.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2338-75-2
Molecular Formula C9H6F3N
Molecular Weight (g/mol) 185.15
MDL Number MFCD00001921
InChI Key QNKOCFJZJWOXDE-UHFFFAOYSA-N
Synonym 4-trifluoromethyl phenylacetonitrile, 2-4-trifluoromethyl phenyl acetonitrile, 4-trifluoromethyl benzyl cyanide, p-trifluoromethylbenzyl cyanide, 4-trifluoromethyl phenyl acetonitrile, 4-trifluoromethyl-phenyl-acetonitrile, benzeneacetonitrile, 4-trifluoromethyl, 4-trifluoromethylbenzyl cyanide, p-trifluoromethylphenylacetonitrile
PubChem CID 75359
IUPAC Name 2-[4-(trifluoromethyl)phenyl]acetonitrile
SMILES FC(F)(F)C1=CC=C(CC#N)C=C1

Specifications

Melting Point 43°C to 47°C
Density 1.23
Boiling Point 131°C to 132°C (20 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
UN Number UN3439
Beilstein 909641
Formula Weight 185.15
Percent Purity 98%
Chemical Name or Material 4-(Trifluoromethyl)phenylacetonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.