missing translation for 'onlineSavingsMsg'
Learn More

[4-(Trifluoromethyl)phenyl]methanesulfonyl chloride, ≥95%, Thermo Scientific™

Product Code. 10120493
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10120493 1 g Amber Glass Bottle 1g
10576504 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10120493 Supplier Thermo Scientific Maybridge Supplier No. MO07768DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 163295-75-8
Molecular Formula C8H6ClF3O2S
Molecular Weight (g/mol) 258.64
MDL Number MFCD01631932
InChI Key KKBNUPMMAGEQAT-UHFFFAOYSA-N
Synonym 4-trifluoromethyl phenyl methanesulfonyl chloride, 4-trifluoromethylbenzylsulfonyl chloride, 4-trifluoromethyl benzylsulfonyl chloride, 4-trifluoromethyl phenyl methyl sulfonyl chloride, 4-chlorosulphonyl methyl benzotrifluoride, 4-trifluoromethyl-alpha-toluenesulfonyl chloride, 4-trifluoromethylphenyl methanesulfonyl chloride, 4-trifluoromethyl-phenyl-methanesulfonyl chloride, alpha-chlorosulphonyl-4-trifluoromethyl toluene
PubChem CID 594332
IUPAC Name [4-(trifluoromethyl)phenyl]methanesulfonyl chloride
SMILES FC(F)(F)C1=CC=C(CS(Cl)(=O)=O)C=C1

Specifications

CAS Min % 95.0
Assay Percent Range 95+%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 258.65
Percent Purity 95%
Chemical Name or Material [4-(Trifluoromethyl)phenyl]methanesulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.