missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)-o-phenylenediamine, 99%, ACROS Organics™

Product Code. 10109941 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10109941

Brand: Thermo Scientific Acros 355300010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 368-71-8
Molecular Formula C7H7F3N2
Molecular Weight (g/mol) 176.142
MDL Number MFCD00042456
InChI Key RQWJHUJJBYMJMN-UHFFFAOYSA-N
Synonym 3,4-diaminobenzotrifluoride, 4-trifluoromethyl benzene-1,2-diamine, 4-trifluoromethyl-1,2-phenylenediamine, 4-trifluoromethyl-o-phenylenediamine, 1,2-benzenediamine, 4-trifluoromethyl, 4-trifluoromethyl-1,2-benzenediamine, 3,4-diamino-benzotrifluoride, 3,4-diamino benzotrifluoride
PubChem CID 520822
IUPAC Name 4-(trifluoromethyl)benzene-1,2-diamine
SMILES C1=CC(=C(C=C1C(F)(F)F)N)N

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 59°C to 62°C
Color Brown-Gray
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 1 g
Formula Weight 176.14
Percent Purity ≥98.5%
Physical Form Powder
Chemical Name or Material 4-(Trifluoromethyl)-o-phenylenediamine
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.