missing translation for 'onlineSavingsMsg'
Learn More

4-Trifluoromethyl-o-phenylenediamine, 98%

Product Code. 11490713
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11490713

Brand: Thermo Scientific Alfa Aesar A12749.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 368-71-8 | C7H7F3N2 | 176.142 g/mol

3,4-Diaminobenzotrifluoride is commonly used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Diaminobenzotrifluoride is commonly used as pharmaceutical intermediate.

Solubility
Miscible with water.

Notes
Stable at room temperature in closed containers under normal storage and handling conditions. Incompatible to oxidizing agents. Store in a cool, dry place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 368-71-8
Molecular Formula C7H7F3N2
Molecular Weight (g/mol) 176.142
MDL Number MFCD00042456
InChI Key RQWJHUJJBYMJMN-UHFFFAOYSA-N
Synonym 3,4-diaminobenzotrifluoride, 4-trifluoromethyl benzene-1,2-diamine, 4-trifluoromethyl-1,2-phenylenediamine, 4-trifluoromethyl-o-phenylenediamine, 1,2-benzenediamine, 4-trifluoromethyl, 4-trifluoromethyl-1,2-benzenediamine, 3,4-diamino-benzotrifluoride, 3,4-diamino benzotrifluoride
PubChem CID 520822
IUPAC Name 4-(trifluoromethyl)benzene-1,2-diamine
SMILES C1=CC(=C(C=C1C(F)(F)F)N)N

Specifications

Melting Point 58°C to 62°C
Quantity 1 g
UN Number UN2811
Beilstein 909258
Solubility Information Miscible with water.
Formula Weight 176.14
Percent Purity 98%
Chemical Name or Material 4-Trifluoromethyl-o-phenylenediamine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.