missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)nicotinonitrile, ≥95%, Thermo Scientific™

Product Code. 10603803
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10603803 1 g 1g
10639505 5 g 5g
10703263 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10603803 Supplier Thermo Scientific Maybridge Supplier No. RF04123DA
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 13600-43-6
Molecular Formula C7H3F3N2
Molecular Weight (g/mol) 172.11
MDL Number MFCD00153107
InChI Key DHIRCRHQLUNYDS-UHFFFAOYSA-N
Synonym 4-trifluoromethyl nicotinonitrile, 3-cyano-4-trifluoromethyl pyridine, 4-trifluoromethyl pyridine-3-carbonitrile, 3-cyano-4-trifluoromethylpyridine, 4-trifluoromethyl-3-pyridinenitrile, 3-pyridinecarbonitrile, 4-trifluoromethyl, 4-trifluoromethyl-nicotinonitrile, pubchem9232, buttpark 4357-45, ksc495c9t
PubChem CID 2736743
IUPAC Name 4-(trifluoromethyl)pyridine-3-carbonitrile
SMILES FC(F)(F)C1=C(C=NC=C1)C#N

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 172.11
Percent Purity 97%
Chemical Name or Material 4-(Trifluoromethyl)nicotinonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.