missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)nicotinic acid, 97%, Thermo Scientific™

Product Code. 10193134
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10193134 5 g Amber Glass Bottle 5g
10160742 250 mg Amber Glass Bottle 250mg
10253322 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10193134 Supplier Thermo Scientific Maybridge Supplier No. RF03948DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 158063-66-2
Molecular Formula C7H4F3NO2
Molecular Weight (g/mol) 191.11
MDL Number MFCD00082626
InChI Key LMRJHNFECNKDKH-UHFFFAOYSA-N
Synonym 4-trifluoromethyl nicotinic acid, 4-trifluoromethyl pyridine-3-carboxylic acid, tfna, 3-pyridinecarboxylic acid, 4-trifluoromethyl, 4-trifluoromethyl-nicotinic acid, 4-trifluoromethylnicotinic acid, 3-carboxy-4-trifluoromethyl pyridine, 4-trifluoromethyl-3-pyridinecarboxylic acid, maybridge1_006267
PubChem CID 2777549
IUPAC Name 4-(trifluoromethyl)pyridine-3-carboxylic acid
SMILES OC(=O)C1=C(C=CN=C1)C(F)(F)F

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 191.11
Percent Purity 97%
Chemical Name or Material 4-(Trifluoromethyl)nicotinic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.