missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)mandelic acid, 98%

Product Code. 11465916
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11465916 5 g 5g
11455916 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11465916 Supplier Thermo Scientific Alfa Aesar Supplier No. B22264.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 395-35-7 | C9H7F3O3 | 220.147 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 395-35-7
Molecular Formula C9H7F3O3
Molecular Weight (g/mol) 220.147
MDL Number MFCD00009724
InChI Key SDGXYUQKJPFLDG-UHFFFAOYSA-N
Synonym 4-trifluoromethyl mandelic acid, 2-hydroxy-2-4-trifluoromethyl phenyl acetic acid, 4-trifluoromethylmandelic acid, hydroxy 4-trifluoromethyl phenyl acetic acid, 4-trifluoromethyl phenylglycolic acid, 2-4-trifluoromethyl phenyl-2-hydroxyacetic acid, p-trifluoromethylmandelic acid
PubChem CID 164593
IUPAC Name 2-hydroxy-2-[4-(trifluoromethyl)phenyl]acetic acid
SMILES C1=CC(=CC=C1C(C(=O)O)O)C(F)(F)F

Specifica

Melting Point 129°C to 131°C
Quantity 5 g
Beilstein 2979949
Formula Weight 220.15
Percent Purity 98%
Chemical Name or Material 4-(Trifluoromethyl)mandelic acid

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.