missing translation for 'onlineSavingsMsg'
Learn More

4-Trifluoromethyl-L-phenylalanine, 95%

Product Code. 15408717
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15408717 250 mg 250mg
15488707 1 g 1g
15498707 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15408717 Supplier Thermo Scientific Alfa Aesar Supplier No. H52170.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 114926-38-4 | C10H10F3NO2 | 233.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
4-fluoro-l-phenylalanine (fF), or 4-trifluoromethyl-l-phenylalanine were introduced inside the chain of peptides to enhances their antimicrobial activity..

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 114926-38-4
Molecular Formula C10H10F3NO2
Molecular Weight (g/mol) 233.19
MDL Number MFCD00044946
InChI Key CRFFPDBJLGAGQL-QMMMGPOBSA-N
Synonym 4-trifluoromethyl-l-phenylalanine, s-2-amino-3-4-trifluoromethyl phenyl propanoic acid, 2s-2-amino-3-4-trifluoromethyl phenyl propanoic acid, l-4-trifluoromethylphenylalanine, l-4-trifluoromethylphe, l-phenylalanine, 4-trifluoromethyl, 4-trifluromethyl-l-phenylalanine hydrochloride, s-2-amino-3-4-trifluoromethyl phenyl propionic acid, h-phe 4-cf3-oh
PubChem CID 2761501
IUPAC Name (2S)-2-amino-3-[4-(trifluoromethyl)phenyl]propanoic acid
SMILES C1=CC(=CC=C1CC(C(=O)O)N)C(F)(F)F

Specifications

Beilstein 5747857
Solubility Information Slightly soluble in water.
Quantity 250 mg
Formula Weight 233.19
Percent Purity 95%
Chemical Name or Material 4-Trifluoromethyl-L-phenylalanine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.