missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)hydrocinnamic Acid, TRC

Product Code. 30449894
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30449894 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30449894 Supplier Toronto Research Chemicals Supplier No. TRCT7965651G
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-(Trifluoromethyl)hydrocinnamic Acid
CAS 53473-36-2
Molecular Formula C10H9F3O2
Quantity 1 g
Synonym 4-(Trifluoromethyl)benzenepropanoic Acid, 3-(4-Trifluoromethylphenyl)propionic Acid, 3-[4-(Trifluoromethyl)phenyl]propanoic acid, 4-(Trifluoromethyl)hydrocinnamic acid, 4-Trifluoromethylbenzenepropanoic acid, p-(Trifluoromethyl)hydrocinnamic acid
InChI Formula InChI=1S/C10H9F3O2/c11-10(12,13)8-4-1-7(2-5-8)3-6-9(14)15/h1-2,4-5H,3,6H2,(H,14,15)
SMILES O=C(CCC1=CC=C(C(F)(F)F)C=C1)O
IUPAC Name 3-(4-(trifluoromethyl)phenyl)propanoic acid
Molecular Weight (g/mol) 218.17
Formula Weight 218.06
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.