missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)cyclohexanone, 97%

Product Code. 15436657
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15436657 1 g 1g
15446657 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15436657 Supplier Thermo Scientific Alfa Aesar Supplier No. H33793.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 75091-99-5 | C7H9F3O | 166.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75091-99-5
Molecular Formula C7H9F3O
Molecular Weight (g/mol) 166.14
MDL Number MFCD00102145
InChI Key IPQDZFUZVJKAKZ-UHFFFAOYSA-N
PubChem CID 2777491
IUPAC Name 4-(trifluoromethyl)cyclohexan-1-one
SMILES FC(F)(F)C1CCC(=O)CC1

Specifications

Boiling Point 67°C to 68°C (16 mmHg)
Quantity 1 g
UN Number UN1224
Synonym 4-trifluoromethyl cyclohexanone, 4-trifluoromethyl cyclohexan-1-one, 4-trifluoromethylcyclohexanone, cyclohexanone, 4-trifluoromethyl, 4-trifluoromethyl-1-cyclohexanone, pubchem16025, 4-trifluoromethyl-cyclohexanone, cyclohexanone,4-trifluoromethyl, 1-oxo-4-trifluoromethyl cyclohexane
Formula Weight 166.14
Percent Purity 97%
Chemical Name or Material 4-(Trifluoromethyl)cyclohexanone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.