missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)cyclohexan-1-one, 97%, Thermo Scientific™

Product Code. 10597444
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10597444 5 g Amber Glass Bottle 5g
10374702 1 g Amber Glass Bottle 1g
10336654 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10597444 Supplier Thermo Scientific Maybridge Supplier No. BTB10356DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 75091-99-5
Molecular Formula C7H9F3O
Molecular Weight (g/mol) 166.14
MDL Number MFCD00102145
InChI Key IPQDZFUZVJKAKZ-UHFFFAOYSA-N
PubChem CID 2777491
IUPAC Name 4-(trifluoromethyl)cyclohexan-1-one
SMILES FC(F)(F)C1CCC(=O)CC1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula CF3C6H9(=O)
Quantity 5 g
Synonym 4-trifluoromethyl cyclohexanone, 4-trifluoromethyl cyclohexan-1-one, 4-trifluoromethylcyclohexanone, cyclohexanone, 4-trifluoromethyl, 4-trifluoromethyl-1-cyclohexanone, pubchem16025, 4-trifluoromethyl-cyclohexanone, cyclohexanone,4-trifluoromethyl, 1-oxo-4-trifluoromethyl cyclohexane
Formula Weight 166.14
Percent Purity 97%
Chemical Name or Material 4-(Trifluoromethyl)cyclohexan-1-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.