missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)benzylamine, 97%

Product Code. 11419767
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11419767 1 g 1g
11429767 5 g 5g
11339449 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11419767 Supplier Thermo Scientific Alfa Aesar Supplier No. B22745.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3300-51-4 | C8H8F3N | 175.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3300-51-4
Molecular Formula C8H8F3N
Molecular Weight (g/mol) 175.15
MDL Number MFCD00010220
InChI Key PRDBLLIPPDOICK-UHFFFAOYSA-N
Synonym 4-trifluoromethyl benzylamine, 4-trifluoromethylbenzylamine, 4-trifluoromethyl phenyl methanamine, 1-4-trifluoromethyl phenyl methanamine, 4-trifluoromethyl benzyl amine, p-trifluoromethylbenzyl amine, 4-trifluoromethyl-benzylamine, p-trifluoromethyl benzylamine, benzenemethanamine, 4-trifluoromethyl
PubChem CID 76804
SMILES NCC1=CC=C(C=C1)C(F)(F)F

Specifications

Boiling Point 80°C to 82°C (15 mmHg)
Flash Point 75°C (167°F)
Quantity 1 g
UN Number UN2735
Beilstein 2640698
IUPAC Name 1-[4-(trifluoromethyl)phenyl]methanamine
Formula Weight 175.15
Percent Purity 98%
Chemical Name or Material 4-(Trifluoromethyl)benzylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.