missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)benzyl Bromide, TRC

Product Code. 30453750
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30453750 5 g 5g
30453243 1 g 1g
30449489 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30453750 Supplier Toronto Research Chemicals Supplier No. TRCT7905855G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 402-49-3
Molecular Formula C8H6BrF3
Molecular Weight (g/mol) 239.03
Synonym alpha'-Bromo-alpha,alpha,alpha-trifluoro-p-xylene, 1-(Bromomethyl)-4-(trifluoromethyl)benzene, 1-Trifluoromethyl-4-(bromomethyl)benzene, 4-Bromomethylbenzotrifluoride, p-(Trifluoromethyl)benzyl bromide, alpha'-Bromo-alpha,alpha,alpha-trifluoro-p-xylene
IUPAC Name 1-(bromomethyl)-4-(trifluoromethyl)benzene
SMILES FC(F)(F)c1ccc(CBr)cc1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C8H6BrF3/c9-5-6-1-3-7(4-2-6)8(10,11)12/h1-4H,5H2
Formula Weight 237.9605
Chemical Name or Material 4-(Trifluoromethyl)benzyl Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.