missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)benzyl alcohol, 98%

Product Code. 11418763
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11418763 100 g 100g
11498753 5 g 5g
11408763 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11418763 Supplier Thermo Scientific Alfa Aesar Supplier No. A10508.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 349-95-1 | C8H7F3O | 176.138 g/mol

4-(Trifluoromethyl)benzyl alcohol, is used as a pharmaceutical intermediate, it is also used to predict the NMR spectrum. 4-(Trifluoromethyl)benzyl alcohol is employed as a reagent in, for example, kinetic studies of phosphonoformate prodrugs and aquachromium(IV).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Trifluoromethyl)benzyl alcohol, is used as a pharmaceutical intermediate, it is also used to predict the NMR spectrum. 4-(Trifluoromethyl)benzyl alcohol is employed as a reagent in, for example, kinetic studies of phosphonoformate prodrugs and aquachromium(IV).

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Keep away from oxidizing agents,reducing agents, acid chlorides, acid anhydrides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 349-95-1
Molecular Formula C8H7F3O
Molecular Weight (g/mol) 176.138
MDL Number MFCD00004661
InChI Key MOOUWXDQAUXZRG-UHFFFAOYSA-N
Synonym 4-trifluoromethyl benzyl alcohol, 4-trifluoromethyl phenyl methanol, p-trifluoromethylbenzyl alcohol, 4-trifluoromethyl benzene methanol, 4-trifluoromethyl benzylic alcohol, benzenemethanol, 4-trifluoromethyl, 4-trifluoromethylbenzyl alcohol, ccris 5117, 4-trifluoromethyl benzylalcohol
PubChem CID 67684
IUPAC Name [4-(trifluoromethyl)phenyl]methanol
SMILES C1=CC(=CC=C1CO)C(F)(F)F

Specifications

Melting Point 18°C to 20°C
Density 1.288
Boiling Point 78°C to 80°C (4 mmHg)
Flash Point 100°C (212°F)
Refractive Index 1.459
Quantity 100 g
Beilstein 1450174
Solubility Information Soluble in water.
Formula Weight 176.14
Percent Purity 98%
Chemical Name or Material 4- (Trifluoromethyl)benzyl alcohol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.