missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)benzoyl chloride, 97%

Product Code. 11442237
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11442237 25 g 25g
11432237 5 g 5g
11452237 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11442237 Supplier Thermo Scientific Alfa Aesar Supplier No. A14176.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 329-15-7 | C8H4ClF3O | 208.56 g/mol

4-(Trifluoromethyl)benzoyl chloride is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Trifluoromethyl)benzoyl chloride is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Store under dry inert gas. Protect from humidity and water. Incompatible with water, bases, alcohol and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 329-15-7
Molecular Formula C8H4ClF3O
Molecular Weight (g/mol) 208.56
MDL Number MFCD00000694
InChI Key OXZYBOLWRXENKT-UHFFFAOYSA-N
Synonym 4-trifluoromethyl benzoyl chloride, p-trifluoromethyl benzoyl chloride, p-trifluoromethylbenzoyl chloride, benzoyl chloride, 4-trifluoromethyl, alpha,alpha,alpha-trifluoro-p-toluoyl chloride, 4-trifluoromethyl benzoylchloride, p-trifluoromethyl benzoic acid chloride, ptf-boc, 4-trifluoromethylbenzoylchloride, 4-trifluoromethyl-benzoyl chloride
PubChem CID 67607
IUPAC Name 4-(trifluoromethyl)benzoyl chloride
SMILES FC(F)(F)C1=CC=C(C=C1)C(Cl)=O

Specifications

Melting Point -3°C
Density 1.404
Boiling Point 188°C to 190°C
Flash Point 78°C (172°F)
Odor Pungent
Refractive Index 1.477
Quantity 25 g
UN Number UN3265
Beilstein 391282
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 208.57
Percent Purity 98%
Chemical Name or Material 4-(Trifluoromethyl)benzoyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.