missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)benzophenone, 98%, Thermo Scientific™

Product Code. 11415666
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11415666 5 g 5g
11425666 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11415666 Supplier Thermo Scientific Alfa Aesar Supplier No. B20775.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 728-86-9
Molecular Formula C14H9F3O
Molecular Weight (g/mol) 250.22
MDL Number MFCD00000400
InChI Key OHTYZZYAMUVKQS-UHFFFAOYSA-N
Synonym 4-trifluoromethyl benzophenone, 4-trifluoromethylbenzophenone, phenyl 4-trifluoromethyl phenyl methanone, p-trifluoromethylbenzophenone, phenyl-4-trifluoromethyl phenyl methanone, methanone, phenyl 4-trifluoromethyl phenyl, fxffr dvr, phenyl 4-trifluoromethyl phenyl ketone, acmc-20amto
PubChem CID 69767
IUPAC Name phenyl-[4-(trifluoromethyl)phenyl]methanone
SMILES FC(F)(F)C1=CC=C(C=C1)C(=O)C1=CC=CC=C1

Specifications

Melting Point 116°C to 119°C
Odor Odorless
Quantity 5 g
Beilstein 1879572
Formula Weight 250.22
Percent Purity 98%
Chemical Name or Material 4-(Trifluoromethyl)benzophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.