missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)benzonitrile, TRC

Product Code. 30454889
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30454889 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30454889 Supplier Toronto Research Chemicals Supplier No. TRCT7917805G
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Chemical Name or Material 4-(Trifluoromethyl)benzonitrile
CAS 455-18-5
Molecular Formula C8H4F3N
Quantity 5 g
Synonym 4-(Trifluoromethyl)benzonitrile, 4-Cyano-α,α,α-trifluorotoluene, 4-Trifluoromethyl-1-cyanobenzene, NSC 88340, p-(Trifluoromethyl)benzonitrile, p-(Trifluoromethyl)cyanobenzene, p-Cyanobenzotrifluoride, α,α,α-Trifluoro-p-tolunitrile
InChI Formula InChI=1S/C8H4F3N/c9-8(10,11)7-3-1-6(5-12)2-4-7/h1-4H
SMILES FC(F)(F)c1ccc(cc1)C#N
IUPAC Name 4-(trifluoromethyl)benzonitrile
Molecular Weight (g/mol) 171.12
Formula Weight 171.0296
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.