missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)benzhydrazide, 98%

Product Code. 11347575
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11347575 1 g 1g
11357575 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11347575 Supplier Thermo Scientific Alfa Aesar Supplier No. L05724.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 339-59-3 | C8H7F3N2O | 204.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 339-59-3
Molecular Formula C8H7F3N2O
Molecular Weight (g/mol) 204.15
MDL Number MFCD00051703
InChI Key GKBDXTNCBPZMFX-UHFFFAOYSA-N
Synonym 4-trifluoromethyl benzhydrazide, 4-trifluoromethyl benzohydrazide, 4-trifluoromethyl benzoic acid hydrazide, 4-trifluoromethylbenzhydrazide, 4-trifluoromethylbenzoic acid hydrazide, 4-trifluoromethyl benzene-1-carbohydrazide, 1-trifluoromethyl benzene-4-carbohydrazide, alpha,alpha,alpha-trifluoro-p-toluic acid hydrazide, benzoic acid, 4-trifluoromethyl-, hydrazide, pubchem4506
PubChem CID 520662
IUPAC Name 4-(trifluoromethyl)benzohydrazide
SMILES NNC(=O)C1=CC=C(C=C1)C(F)(F)F

Specifications

Melting Point 117°C to 121°C
Quantity 1 g
Beilstein 1968848
Formula Weight 204.15
Percent Purity 98%
Chemical Name or Material 4-(Trifluoromethyl)benzhydrazide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.