missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Trifluoromethyl)benzene-1,2-diamine, 97%, Maybridge
Click to view available options
:
Amber Glass Bottle; 1g
Specifications
Specifications
| CAS | 368-71-8 |
| CAS Min % | 94.5 |
| CAS Max % | 100.0 |
| Assay Percent Range | 95% |
| Packaging | Amber Glass Bottle |
| Molecular Formula | C7H7F3N2 |
| MDL Number | MFCD00042456 |
| Quantity | 1 g |
| Synonym | 3,4-diaminobenzotrifluoride, 4-trifluoromethyl benzene-1,2-diamine, 4-trifluoromethyl-1,2-phenylenediamine, 4-trifluoromethyl-o-phenylenediamine, 1,2-benzenediamine, 4-trifluoromethyl, 4-trifluoromethyl-1,2-benzenediamine, 3,4-diamino-benzotrifluoride, 3,4-diamino benzotrifluoride |
| InChI Key | RQWJHUJJBYMJMN-UHFFFAOYSA-N |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction