missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Trifluoromethyl)aniline, ≥97%, Thermo Scientific™
Specifications
Specifications
| CAS Min % | 97.0 |
| Assay Percent Range | 97+% |
| Linear Formula | H2NC6H4CF3 |
| Quantity | 25 g |
| Formula Weight | 161.13 |
| Percent Purity | 97% |
| Chemical Name or Material | 4-(Trifluoromethyl)aniline |
Safety and Handling
Product Identifier
- 4-Aminobenzotrifluoride
- Danger
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 1
- H301-Toxic if swallowed.
- H318-Causes serious eye damage.
- H331-Toxic if inhaled.
- H372-Causes damage to organs.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contain: 4-Aminobenzotrifluoride
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?