missing translation for 'onlineSavingsMsg'
Learn More

4'-(Trifluoromethyl)acetophenone, 98+%

Product Code. 11492947
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
25 g
100 g
missing translation for 'unitSize':
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity missing translation for 'unitSize'
11492947 5 g 5g
11402957 25 g 25g
11412957 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 11492947 missing translation for 'mfr' Thermo Scientific Alfa Aesar missing translation for 'supplierNo' A15529.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 709-63-7 | C9H7F3O | 188.149 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 709-63-7
Molecular Formula C9H7F3O
Molecular Weight (g/mol) 188.149
MDL Number MFCD00000401
InChI Key HHAISVSEJFEWBZ-UHFFFAOYSA-N
Synonym 4'-trifluoromethyl acetophenone, 1-4-trifluoromethyl phenyl ethanone, 1-4-trifluoromethyl phenyl ethan-1-one, 4-trifluoromethylacetophenone, 4-acetylbenzotrifluoride, 4-trifluoromethyl acetophenone, p-trifluoromethylacetophenone, 4'-trifluoromethylacetophenone, ethanone, 1-4-trifluoromethyl phenyl
PubChem CID 69731
IUPAC Name 1-[4-(trifluoromethyl)phenyl]ethanone
SMILES CC(=O)C1=CC=C(C=C1)C(F)(F)F

Specifications

Melting Point 29°C to 35°C
Density 0.924
Boiling Point 83°C to 85°C (9 mmHg)
Flash Point 84°C (183°F)
Odor Characteristic
Refractive Index 1.462
Quantity 5 g
Beilstein 1870425
Formula Weight 188.15
Percent Purity ≥98%
Chemical Name or Material 4'-(Trifluoromethyl)acetophenone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.