missing translation for 'onlineSavingsMsg'
Learn More

4-Trifluoromethyl-1H-pyrazole, 97%

Product Code. 15427337
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
15427337 250 mg 250mg
15417337 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 15427337

Brand: Thermo Scientific Alfa Aesar H34396.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 52222-73-8 | C4H3F3N2 | 136.077 g/mol

It is used as pharmaceutical intermediates. The palladium-catalyzed cross-coupling reactions of 5-tributylstannyl-4-fluoro-1H-pyrazole with aryl iodides provided high yields of the corresponding 5-aryl-4-fluoro-1H-pyrazoles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediates. The palladium-catalyzed cross-coupling reactions of 5-tributylstannyl-4-fluoro-1H-pyrazole with aryl iodides provided high yields of the corresponding 5-aryl-4-fluoro-1H-pyrazoles.

Notes
Incompatible with strong oxidizing agents. Store at room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52222-73-8
Molecular Formula C4H3F3N2
Molecular Weight (g/mol) 136.077
MDL Number MFCD11226572
InChI Key KDEJQUNODYXYBJ-UHFFFAOYSA-N
Synonym 4-trifluoromethyl-1h-pyrazole, 1h-pyrazole, 4-trifluoromethyl, 4-trifluoromethylpyrazole, 4-trifluoromethyl pyrazole, 4-trifluoromethyl-1 h-pyrazole, 4-trifluoromethyl ;-1h-pyrazole
PubChem CID 12777795
IUPAC Name 4-(trifluoromethyl)-1H-pyrazole
SMILES C1=C(C=NN1)C(F)(F)F

Specifications

Melting Point 76°C to 78°C
Quantity 250 mg
Formula Weight 136.1
Percent Purity 97%
Chemical Name or Material 4-Trifluoromethyl-1H-pyrazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.