missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)-1-indanone, 97%, Thermo Scientific Chemicals

Product Code. 15415398
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15415398 1 g 1g
15425398 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15415398 Supplier Thermo Scientific Alfa Aesar Supplier No. H59165.03
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 68755-42-0 | Molecular Formula: C10H7F3O | Molecular Weight: 200.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 68755-42-0
Molecular Formula C10H7F3O
Molecular Weight (g/mol) 200.16
MDL Number MFCD07772121
InChI Key LJVBFMQEZSEGRL-UHFFFAOYSA-N
Synonym 4-trifluoromethyl-1-indanone, 4-trifluoromethyl-2,3-dihydro-1h-inden-1-one, 4-trifluoromethyl-2,3-dihydroinden-1-one, 1h-inden-1-one, 2,3-dihydro-4-trifluoromethyl, 4-trifluoromethyl indan-1-one, 2,3-dihydro-4-trifluoromethyl-1h-inden-1-one, 2,3-dihydro-1-oxo-4-trifluoromethyl-1h-indene, acmc-1b8x3, 2,3-dihydro-4-trifluoromethyl-1h-inden-1-one
PubChem CID 21921093
IUPAC Name 4-(trifluoromethyl)-2,3-dihydroinden-1-one
SMILES C1CC(=O)C2=C1C(=CC=C2)C(F)(F)F

Specifica

Melting Point 38°C to 42°C
Boiling Point 67°C to 72°C (0.1 mmHg)
Flash Point >108°C (226°F)
Quantity 1 g
Formula Weight 200.16
Percent Purity 97%
Chemical Name or Material 4-(Trifluoromethyl)-1-indanone

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.