missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethoxy)phenylacetic acid, 98%

Product Code. 11494077
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11494077 5 g 5g
11484077 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11494077 Supplier Thermo Scientific Alfa Aesar Supplier No. A19512.06
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 4315-07-5 | C9H6F3O3 | 219.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4315-07-5
MDL Number MFCD00061239

Specifications

Melting Point 86°C to 88°C
Molecular Formula C9H6F3O3
Quantity 5 g
Beilstein 2695336
Synonym 4-trifluoromethoxy phenylacetic acid, 2-4-trifluoromethoxy phenyl acetic acid, 4-trifluoromethoxy phenyl acetic acid, 4-trifluoromethoxy phenylaceticacid, 4-trifluoromethoxy-phenyl-acetic acid, 4-trifluoromethoxyphenylacetic acid, benzeneacetic acid, 4-trifluoromethoxy, maybridge3_006163
InChI Key ZMTIBXQPXBUWPQ-UHFFFAOYSA-M
SMILES C1=CC(=CC=C1CC(=O)O)OC(F)(F)F
IUPAC Name 2-[4-(trifluoromethoxy)phenyl]acetic acid
Molecular Weight (g/mol) 219.14
PubChem CID 2777319
Formula Weight 220.15
Percent Purity 98%
Chemical Name or Material 4-(Trifluoromethoxy)phenylacetic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.