missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethoxy)phenyl isocyanate, 97%, Thermo Scientific™

Product Code. 10235464
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10235464 1 g 1g
10295504 10 g 10g
10001924 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10235464 Supplier Thermo Scientific Maybridge Supplier No. SB01053DA
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 35037-73-1
Molecular Formula C8H4F3NO2
Molecular Weight (g/mol) 203.12
MDL Number MFCD00035702
InChI Key LGPKFIGMLPDYEA-UHFFFAOYSA-N
Synonym 4-trifluoromethoxy phenyl isocyanate, 1-isocyanato-4-trifluoromethoxy benzene, 4-trifluoromethoxy phenylisocyanate, 4-trifluoromethoxyphenylisocyanate, benzene, 1-isocyanato-4-trifluoromethoxy, 1-isocyanato-4-trifluoromethoxy-benzene, p-trifluoromethoxy phenyl isocyanate, isocyanic acid 4-trifluoromethoxy phenyl ester, 4-trifluoromethoxy benzenisocyanate
PubChem CID 92298
IUPAC Name 1-isocyanato-4-(trifluoromethoxy)benzene
SMILES C1=CC(=CC=C1N=C=O)OC(F)(F)F

Specifications

CAS Min % 97.0
Density 1.34g/mL
Assay Percent Range 97%
Packaging Amber glass bottle
Linear Formula CF3OC6H4NCO
Quantity 1 g
Formula Weight 203.12
Percent Purity 97%
Chemical Name or Material 4-(Trifluoromethoxy)phenyl isocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.