missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethoxy)phenyl isocyanate, 97%, Thermo Scientific™

Click to view available options
Quantity:
1g
10g
25g
This item is not returnable. View return policy
35037-73-1
C8H4F3NO2
203.12
MFCD00035702
LGPKFIGMLPDYEA-UHFFFAOYSA-N
4-trifluoromethoxy phenyl isocyanate, 1-isocyanato-4-trifluoromethoxy benzene, 4-trifluoromethoxy phenylisocyanate, 4-trifluoromethoxyphenylisocyanate, benzene, 1-isocyanato-4-trifluoromethoxy, 1-isocyanato-4-trifluoromethoxy-benzene, p-trifluoromethoxy phenyl isocyanate, isocyanic acid 4-trifluoromethoxy phenyl ester, 4-trifluoromethoxy benzenisocyanate
92298
1-isocyanato-4-(trifluoromethoxy)benzene
C1=CC(=CC=C1N=C=O)OC(F)(F)F
This item is not returnable. View return policy
35037-73-1
C8H4F3NO2
203.12
MFCD00035702
LGPKFIGMLPDYEA-UHFFFAOYSA-N
4-trifluoromethoxy phenyl isocyanate, 1-isocyanato-4-trifluoromethoxy benzene, 4-trifluoromethoxy phenylisocyanate, 4-trifluoromethoxyphenylisocyanate, benzene, 1-isocyanato-4-trifluoromethoxy, 1-isocyanato-4-trifluoromethoxy-benzene, p-trifluoromethoxy phenyl isocyanate, isocyanic acid 4-trifluoromethoxy phenyl ester, 4-trifluoromethoxy benzenisocyanate
92298
1-isocyanato-4-(trifluoromethoxy)benzene
C1=CC(=CC=C1N=C=O)OC(F)(F)F
CAS Min % 97.0
Density 1.34g/mL
Assay Percent Range 97%
Linear Formula CF3OC6H4NCO
Quantity 25g
Formula Weight 203.12
Percent Purity 97%
Chemical Name or Material 4-(Trifluoromethoxy)phenyl isocyanate