missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethoxy)benzylamine, 97%

Product Code. 10277460 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10277460 1 g Glass bottle 1g
10456042 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10277460 Supplier Thermo Scientific Acros Supplier No. 297700010
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 93919-56-3 | C8H8F3NO | 191.153 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93919-56-3
MDL Number MFCD00061237
InChI Key DBGROTRFYBSUTR-UHFFFAOYSA-N
Synonym 4-trifluoromethoxy benzylamine, 4-trifluoromethoxy phenyl methanamine, 1-4-trifluoromethoxy phenyl methanamine, 4-trifluoromethoxybenzylamine, benzenemethanamine, 4-trifluoromethoxy, p-trifluoromethoxybenzylamine, p-trifluoromethoxy benzylamine, 4-trifluoromethoxy benzyl amine, 4-trifluoromethoxy phenyl methylamine
PubChem CID 571846
IUPAC Name [4-(trifluoromethoxy)phenyl]methanamine
SMILES C1=CC(=CC=C1CN)OC(F)(F)F

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.25g/mL
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C8H8F3NO
Linear Formula CF3OC6H4CH2NH2
Refractive Index 1.4510 to 1.4530 (20°C, 589nm)
Quantity 1 g
Molecular Weight (g/mol) 191.153
Formula Weight 191.15
Percent Purity ≥96.0%
Physical Form Liquid
Chemical Name or Material 4-(Trifluoromethoxy)benzylamine, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 4-(Trifluoromethoxy)benzylamine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.