missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethoxy)benzyl bromide, 98%, Thermo Scientific™

Product Code. 10297070 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mg
2.5 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
2.5g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10297070 2.5 g Glass bottle 2.5g
10712695 500 mg Glass bottle 500mg
10702695 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10297070 Supplier Thermo Scientific Acros Supplier No. 293510025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 50824-05-0
Molecular Formula C8H6BrF3O
Molecular Weight (g/mol) 255.03
MDL Number MFCD00061238
InChI Key JDNPUJCKXLOHOW-UHFFFAOYSA-N
Synonym 4-trifluoromethoxy benzyl bromide, 1-bromomethyl-4-trifluoromethoxy benzene, 4-trifluoromethoxybenzyl bromide, 4-trifluoromethoxy benzylbromide, p-trifluoromethoxy benzyl bromide, alpha-bromo-4-trifluoromethoxy toluene, benzene, 1-bromomethyl-4-trifluoromethoxy, 4-trifluoromethoxy-benzyl bromide, 4-bromomethyl phenoxy trifluoromethane, 1-bromomethyl-4-trifluoromethoxy-benzene
PubChem CID 142785
IUPAC Name 1-(bromomethyl)-4-(trifluoromethoxy)benzene
SMILES C1=CC(=CC=C1CBr)OC(F)(F)F

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 23°C to 24°C
Color Yellow
Density 1.5800g/mL
Boiling Point 82°C to 84°C (10.0 mmHg)
Flash Point 94°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CF3OC6H4CH2Br
Refractive Index 1.4800 to 1.482
Quantity 2.5 g
Specific Gravity 1.58
Formula Weight 255.03
Percent Purity 98%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material 4-(Trifluoromethoxy)benzyl bromide
Show More Show Less
compliance-icons
Product Identifier
  • 4-(Trifluoromethoxy)benzyl bromide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.