missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Trifluoromethoxy)benzyl Alcohol, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | 4-(Trifluoromethoxy)benzyl Alcohol |
| CAS | 1736-74-9 |
| Molecular Formula | C8H7F3O2 |
| Quantity | 1 g |
| Synonym | 4-(Trifluoromethoxy)-benzenemethanol, p-(Trifluoromethoxy)-benzyl Alcohol, 4-(Trifluoromethoxy)benzenemethanol, [4-(Trifluoromethoxy)phenyl]methanol |
| InChI Formula | InChI=1S/C8H7F3O2/c9-8(10,11)13-7-3-1-6(5-12)2-4-7/h1-4,12H,5H2 |
| SMILES | OCc1ccc(OC(F)(F)F)cc1 |
| IUPAC Name | [4-(trifluoromethoxy)phenyl]methanol |
| Molecular Weight (g/mol) | 192.14 |
| Formula Weight | 192.0398 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction