missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethoxy)benzoyl chloride, 98%

Product Code. 10591901 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10591901 1 g Glass bottle 1g
10434545 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10591901 Supplier Thermo Scientific Acros Supplier No. 306350010
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 36823-88-8 | C8H4ClF3O2 | 224.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 36823-88-8
Molecular Formula C8H4ClF3O2
Molecular Weight (g/mol) 224.56
MDL Number MFCD00052329
InChI Key ZXKKOFJYPRJFIE-UHFFFAOYSA-N
Synonym 4-trifluoromethoxy benzoyl chloride, p-trifluoromethoxybenzoyl chloride, benzoyl chloride, 4-trifluoromethoxy, 4-trifluoromethoxy benzene-1-carbonyl chloride, 4-trifluoromethoxy benzoylchloride, 4-trifluoromethoxy-benzoyl chloride, p-trifluoromethoxy benzoyl chloride, acmc-209ip1, ksc495i1f, p-trifluoromethoxybenzoylchloride
PubChem CID 142110
IUPAC Name 4-(trifluoromethoxy)benzoyl chloride
SMILES C1=CC(=CC=C1C(=O)Cl)OC(F)(F)F

Especificaciones

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Pink
Density 1.4200g/mL
Boiling Point 55°C to 56°C (2.0 mmHg)
Flash Point 32°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CF3OC6H4COCl
Refractive Index 1.4710 to 1.473
Quantity 1 g
Specific Gravity 1.42
Formula Weight 224.56
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-(Trifluoromethoxy)benzoyl chloride
Mostrar más Mostrar menos

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.