missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethoxy)benzoyl chloride, 97%, Thermo Scientific™

Product Code. 11444037
Change view
Click to view available options
Quantity:
1 g
25 g
Unit Size:
1g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11444037 25 g 25g
11424037 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11444037 Supplier Thermo Scientific Alfa Aesar Supplier No. A19452.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36823-88-8
Molecular Formula C8H4ClF3O2
Molecular Weight (g/mol) 224.563
MDL Number MFCD00052329
InChI Key ZXKKOFJYPRJFIE-UHFFFAOYSA-N
Synonym 4-trifluoromethoxy benzoyl chloride, p-trifluoromethoxybenzoyl chloride, benzoyl chloride, 4-trifluoromethoxy, 4-trifluoromethoxy benzene-1-carbonyl chloride, 4-trifluoromethoxy benzoylchloride, 4-trifluoromethoxy-benzoyl chloride, p-trifluoromethoxy benzoyl chloride, acmc-209ip1, ksc495i1f, p-trifluoromethoxybenzoylchloride
PubChem CID 142110
IUPAC Name 4-(trifluoromethoxy)benzoyl chloride
SMILES C1=CC(=CC=C1C(=O)Cl)OC(F)(F)F

Specifications

Density 1.425
Boiling Point 55°C to 56°C (2 mmHg)
Flash Point 90°C (194°F)
Refractive Index 1.472
Quantity 25 g
UN Number UN3265
Beilstein 2723543
Sensitivity Moisture sensitive
Formula Weight 224.57
Percent Purity 97%
Chemical Name or Material 4-(Trifluoromethoxy)benzoyl chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.