missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethoxy)benzenesulfonamide, 97%, Thermo Scientific™

Catalog Number 11345555
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1513-45-7
C7H6F3NO3S
241.184
MFCD00798381
RGOJCHYYBKMRLL-UHFFFAOYSA-N
4-trifluoromethoxy benzenesulfonamide, p-trifluoromethoxy benzenesulfonamide, 4-trifluoromethoxy benzenesulphonamide, 4-trifluoromethoxy benzene-1-sulfonamide, benzenesulfonamide, 4-trifluoromethoxy, pubchem11787, rgojchyybkmrll-uhfffaoysa, 4-trifluoromethoxybenzensulfonamide, trifluoromethoxy benzenesulfonamide, 4-trifluoromethoxybenzenesulphonyl amine
2777211
4-(trifluoromethoxy)benzenesulfonamide
C1=CC(=CC=C1OC(F)(F)F)S(=O)(=O)N
This item is not returnable. View return policy
1513-45-7
C7H6F3NO3S
241.184
MFCD00798381
RGOJCHYYBKMRLL-UHFFFAOYSA-N
4-trifluoromethoxy benzenesulfonamide, p-trifluoromethoxy benzenesulfonamide, 4-trifluoromethoxy benzenesulphonamide, 4-trifluoromethoxy benzene-1-sulfonamide, benzenesulfonamide, 4-trifluoromethoxy, pubchem11787, rgojchyybkmrll-uhfffaoysa, 4-trifluoromethoxybenzensulfonamide, trifluoromethoxy benzenesulfonamide, 4-trifluoromethoxybenzenesulphonyl amine
2777211
4-(trifluoromethoxy)benzenesulfonamide
C1=CC(=CC=C1OC(F)(F)F)S(=O)(=O)N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 146°C to 148°C
Quantity 1 g
Beilstein 2653449
Formula Weight 241.19
Percent Purity 97%
Chemical Name or Material 4-(Trifluoromethoxy)benzenesulfonamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'