missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethoxy)benzaldehyde, 96%

Product Code. 11473424
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11473424 5 g 5g
11483424 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11473424 Supplier Thermo Scientific Alfa Aesar Supplier No. A10671.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 659-28-9 | C8H5F3O2 | 190.121 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 659-28-9
Molecular Formula C8H5F3O2
Molecular Weight (g/mol) 190.121
MDL Number MFCD00041530
InChI Key XQNVDQZWOBPLQZ-UHFFFAOYSA-N
Synonym 4-trifluoromethoxy benzaldehyde, p-trifluoromethoxybenzaldehyde, p-trifluoromethoxy benzaldehyde, benzaldehyde, 4-trifluoromethoxy, 4-trifluoromethoxy-benzaldehyde, alpha,alpha,alpha-trifluoroanisaldehyde, pubchem1471, acmc-209nsx
PubChem CID 69573
IUPAC Name 4-(trifluoromethoxy)benzaldehyde
SMILES C1=CC(=CC=C1C=O)OC(F)(F)F

Specifications

Density 1.331
Boiling Point 93°C (27 mmHg)
Flash Point 71°C (159°F)
Refractive Index 1.458
Quantity 5 g
Beilstein 1949135
Sensitivity Air Sensitive
Formula Weight 190.12
Percent Purity 96%
Chemical Name or Material 4- (Trifluoromethoxy)benzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.