missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethoxy)anisole, 98+%

Product Code. 11438627
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11438627 5 g 5g
11448627 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11438627 Supplier Thermo Scientific Alfa Aesar Supplier No. A19746.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 710-18-9 | C8H7F3O2 | 192.137 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 710-18-9
Molecular Formula C8H7F3O2
Molecular Weight (g/mol) 192.137
MDL Number MFCD00216942
InChI Key NOAFZIOGGDPYKK-UHFFFAOYSA-N
Synonym 4-trifluoromethoxy anisole, 1-methoxy-4-trifluoromethoxy benzene, p-trifluoromethoxy anisole, 4-trifluoromethoxyanisole, benzene, 1-methoxy-4-trifluoromethoxy, 4-methoxy trifluoromethoxy benzene, hydroquinone methyl trifluoromethyl ether, trifluoro 4-methoxyphenoxy methane, pubchem3570, acmc-1bcz3
PubChem CID 2724973
IUPAC Name 1-methoxy-4-(trifluoromethoxy)benzene
SMILES COC1=CC=C(C=C1)OC(F)(F)F

Specifications

Density 1.272
Boiling Point 163°C to 165°C
Flash Point 58°C (136°F)
Refractive Index 1.432
Quantity 5 g
UN Number UN1993
Beilstein 2556809
Formula Weight 192.14
Percent Purity ≥98%
Chemical Name or Material 4-(Trifluoromethoxy)anisole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.