missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethoxy)aniline, TRC

Product Code. 30132861
Change view
Click to view available options
Quantity:
50 g
100 g
250 g
Unit Size:
100g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132861 100 g 100g
30135856 250 g 250g
30006378 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132861 Supplier Toronto Research Chemicals Supplier No. TRCT790700100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 461-82-5
Molecular Formula C7 H6 F3 N O
Molecular Weight (g/mol) 177.12
Synonym 4-(Trifluoromethoxy)-benzenamine, p-Anisidine, α,α,α-trifluoro- (6CI,7CI,8CI), (4-(Trifluoromethoxy)phenyl)amine, 1-Amino-4-(trifluoromethoxy)benzene, 4-(Trifluoromethoxy)aniline, 4-(Trifluoromethoxy)benzenamine, 4-Trifluoromethoxybenzeneamine, 4-[(Trifluoromethyl)oxy]aniline, p-(Trifluoromethoxy)aniline, p-Trifluoromethoxyphenylamine, α,α,α-Trifluoro-p-anisidine
IUPAC Name 4-(trifluoromethoxy)aniline
SMILES Nc1ccc(OC(F)(F)F)cc1

Specifications

Quantity 100 g
InChI Formula InChI=1S/C7H6F3NO/c8-7(9,10)12-6-3-1-5(11)2-4-6/h1-4H,11H2
Formula Weight 177.0401
Chemical Name or Material 4-(Trifluoromethoxy)aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.