missing translation for 'onlineSavingsMsg'
Learn More

4-trans-Hydroxy-L-proline ethyl ester hydrochloride, 97%, Thermo Scientific™

Catalog Number p-7056611
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
33996-30-4
C7H14ClNO3
195.643
MFCD00040589
HHXSZDXMSRXWJV-IBTYICNHSA-N
h-hyp-oet hcl, h-hyp-oet.hcl, 2s,4r-ethyl 4-hydroxypyrrolidine-2-carboxylate hydrochloride, l-4-hydroxyproline ethyl ester hydrochloride, ethyl trans-4-hydroxy-l-prolinate hydrochloride, trans-4-hydroxy-l-proline ethyl ester hydrochloride, ethyl 2s,4r-4-hydroxypyrrolidine-2-carboxylate hydrochloride, ethyl trans-4-hydroxy-l-prolinate hcl, 2s,4r---ethyl 4-hydroxy-2-pyrrolidincarboxylate hydrochloride 99+%, 2s,4r-4-hydroxyproline ethyl ester hydrochloride
12709304
ethyl (2S,4R)-4-hydroxypyrrolidine-2-carboxylate;hydrochloride
CCOC(=O)C1CC(CN1)O.Cl
This item is not returnable. View return policy
33996-30-4
C7H14ClNO3
195.643
MFCD00040589
HHXSZDXMSRXWJV-IBTYICNHSA-N
h-hyp-oet hcl, h-hyp-oet.hcl, 2s,4r-ethyl 4-hydroxypyrrolidine-2-carboxylate hydrochloride, l-4-hydroxyproline ethyl ester hydrochloride, ethyl trans-4-hydroxy-l-prolinate hydrochloride, trans-4-hydroxy-l-proline ethyl ester hydrochloride, ethyl 2s,4r-4-hydroxypyrrolidine-2-carboxylate hydrochloride, ethyl trans-4-hydroxy-l-prolinate hcl, 2s,4r---ethyl 4-hydroxy-2-pyrrolidincarboxylate hydrochloride 99+%, 2s,4r-4-hydroxyproline ethyl ester hydrochloride
12709304
ethyl (2S,4R)-4-hydroxypyrrolidine-2-carboxylate;hydrochloride
CCOC(=O)C1CC(CN1)O.Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color White
Beilstein 3656870
Formula Weight 195.64
Percent Purity 97%
Quantity 25 g
Physical Form Powder
Chemical Name or Material 4-trans-Hydroxy-L-proline ethyl ester hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'