missing translation for 'onlineSavingsMsg'
Learn More

4-(Thien-3-yl)benzoic acid, 97%, Thermo Scientific™

Product Code. 15229686 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15229686 5 g 5g
15219686 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15229686 Supplier Thermo Scientific Acros Supplier No. 450420050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 29886-64-4
Molecular Formula C11H8O2S
Molecular Weight (g/mol) 204.25
InChI Key FISAUHGRILVMDP-UHFFFAOYSA-N
Synonym 4-3-thienyl benzoic acid, 4-thiophen-3-yl benzoic acid, 4-thiophen-3-yl-benzoic acid, benzoic acid, 4-3-thienyl, 4-thien-3-ylbenzoic acid, 4-3-thienyl-benzoic acid, maybridge3_004545, acmc-1cisg
PubChem CID 736862
IUPAC Name 4-thiophen-3-ylbenzoic acid
SMILES C1=CC(=CC=C1C2=CSC=C2)C(=O)O

Specifications

Melting Point 281.0°C to 283.0°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Quantity 5 g
Formula Weight 204.25
Percent Purity 97%
Chemical Name or Material 4-(Thien-3-yl)benzoic acid
compliance-icons
Product Identifier
  • 4-(Thien-3-yl)benzoic acid, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P264-Wash face, hands and any thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.