missing translation for 'onlineSavingsMsg'
Learn More

(4-thien-2-ylphenyl)methanol, Thermo Scientific™

Product Code. 10112884
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber glass bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10112884 5 g Amber glass bottle 5g
10630315 250 mg Amber glass bottle 250mg
10493013 1 g Amber glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10112884 Supplier Thermo Scientific Maybridge Supplier No. CC12009DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 81443-44-9
Molecular Formula C11H10OS
Molecular Weight (g/mol) 190.26
InChI Key KHZSWSHUIYOMSH-UHFFFAOYSA-N
Synonym 4-thien-2-ylphenyl methanol, 4-thiophen-2-yl phenyl methanol, thiophene,2-4-methylphenyl, benzenemethanol, 4-2-thienyl, 4-thien-2-yl-phenyl methanol, pubchem24308, 4-2-thienyl benzyl alcohol, 4-thien-2-yl benzyl alcohol, 4-2-thienyl phenyl methanol
PubChem CID 2795254
IUPAC Name (4-thiophen-2-ylphenyl)methanol
SMILES C1=CSC(=C1)C2=CC=C(C=C2)CO

Specifications

Packaging Amber glass bottle
Quantity 5 g
Formula Weight 190.26
Percent Purity 97%
Chemical Name or Material (4-thien-2-ylphenyl)methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.