missing translation for 'onlineSavingsMsg'
Learn More

4-(thien-2-ylmethyl)benzylamine hydrochloride, 97%, Thermo Scientific™

Product Code. 10256004
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10256004 1 g Amber Glass Bottle 1g
10595184 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10256004 Supplier Thermo Scientific Maybridge Supplier No. CC70663DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 1112459-82-1
Molecular Formula C12H14ClNS
Molecular Weight (g/mol) 239.761
MDL Number MFCD12198118
InChI Key BRDMSBXGZSSVJT-UHFFFAOYSA-N
Synonym 4-thien-2-ylmethyl benzylamine hydrochloride, 4-thiophen-2-ylmethyl phenyl methanamine hydrochloride, 4-2-thienylmethyl phenyl methylamine, chloride, 4-thien-2-ylmethyl phenyl methylamine hydrochloride, 1-4-thiophen-2-ylmethyl phenyl methanamine hydrochloride, 1-4-thiophen-2-yl methyl phenyl methanamine-hydrogen chloride 1/1
PubChem CID 43811053
IUPAC Name [4-(thiophen-2-ylmethyl)phenyl]methanamine;hydrochloride
SMILES C1=CSC(=C1)CC2=CC=C(C=C2)CN.Cl

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 239.77
Percent Purity 97%
Chemical Name or Material 4-(Thien-2-ylmethyl)benzylamine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.