missing translation for 'onlineSavingsMsg'
Learn More

4-Tetrahydropyranyl methanesulfonate, 95%

Product Code. 15446927
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15446927

Brand: Thermo Scientific Alfa Aesar H34026.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 134419-59-3 | C6H12O4S | 180.218 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents, acids and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 134419-59-3
Molecular Formula C6H12O4S
Molecular Weight (g/mol) 180.218
MDL Number MFCD11505063
InChI Key GSEZHCLWHDZJAB-UHFFFAOYSA-N
Synonym tetrahydro-2h-pyran-4-yl methanesulfonate, 2h-pyran-4-ol, tetrahydro-, methanesulfonate, acmc-20mvcv, 4-oxanyl methanesulfonate, tetrahydropyran-4-ol methane sulfonate, tetrahydro-2h-pyran-4-ol methanesulfonate, tetrahydro-2h-pyran-4-yl methanesulphonate, methansulfonic acid tetrahydropyran-4-yl ester, methanesulfonic acid tetrahydropyran-4-yl ester
PubChem CID 11344352
IUPAC Name oxan-4-yl methanesulfonate
SMILES CS(=O)(=O)OC1CCOCC1

Specifications

Quantity 5 g
Formula Weight 180.22
Percent Purity 95%
Chemical Name or Material 4-Tetrahydropyranyl methanesulfonate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.